You can import a protein or peptide sequence into the Theoretical Protein/Peptide Manager from a FASTA file or manually enter it.
Open the Theoretical Protein/Peptide Manager
Procedure
- Open the Sequence Manager by selecting Sequence Manager on the BioPharma Finder home page.
- Select the Theoretical Protein/Peptide Manager tab.
- The Theoretical Protein/Peptide Manager page opens, showing the Sequence area and parameters.
Theoretical Protein/Peptide Manager page
Import a protein or peptide sequence from a FASTA file
Procedure
- In the Theoretical Protein/Peptide Manager, select Import Protein Sequence on the command bar and browse to the folder containing the FASTA file.
- The dialog box displays all of the FASTA files in the selected folder.
- The FASTA file must have the ".fasta" extension for the application to be able to find the file.
- Select a FASTA file to import and then select Open.
- The application displays the sequence information from the FASTA file in the Enter Peptide or Protein Sequence box, overwriting any previous content in this box.
- You can import FASTA files of 1 MB or less.
- If a FASTA file contains invalid amino acids or an invalid format, an error message appears. If an error message appears during the import operation, open the FASTA file in a text editor such as Notepad and verify the format of each chain.
Manually create a protein or peptide sequence
Procedure
- In the Sequence area of the Theoretical Protein/Peptide Manager, type the protein or peptide sequence in the Enter Peptide or Protein Sequence box.
NOTE
You cannot specify both proteins and peptides at the same time; however, you can enter them in two separate sessions.
- When inputting protein sequence information in the Enter Peptide or Protein Sequence box, you must:
- Use the approved amino acid alphabet.
- See Amino acid letter codes.
- Begin each chain with a single-line description.
- Use a greater-than (>) sign at the start of this line to distinguish the chain description from the sequence data.
- Use no space between the > and the first letter of the identifier in the description.
- Follow the description with lines of sequence data in the form of amino acid single-letter codes.
- A chain ends when another line starting with > appears, indicating the start of another chain.
- When inputting peptide sequence information in the Enter Peptide or Protein Sequence box, you must:
- Use the approved amino acid alphabet.
- See Amino acid letter codes.
- Use a contiguous sequence of characters to define each peptide.
- Enter multiple peptides on separate lines.
- If you want to perform digestion on the protein, select the Perform Digestion checkbox at the top right of the Sequence area.
- For more information on digestion parameters for theoretical proteins, see Specify digestion parameters.
- To perform the theoretical experiment without changing the default Digestion Parameters (optional), Add/Edit Target m/z, or Add/Edit Modifications, select Process.
- The Results table displays the results of the theoretical experiment.
- See Results table parameters.
- Otherwise, you can edit the Digestion Parameters, Add/Edit Target m/z, or Add/Edit Modifications as needed.
- See the following topics:
- •Specify digestion parameters
- • Add/edit target m/z parameters
- •Add/edit modifications