Delete an HCP database - BioPharma Finder Software - Delete an HCP database - BioPharma Finder Software - BioPharma Finder Software
BioPharma Finder 5.2 Intact Mass Analysis User Guide
- Product Tree
- Ardia Platform > Software > BioPharma Finder Software
- Desktop Software > BioPharma Finder Software
- Document Type
- User Guide
- Software version
- 5.2
- Open the Sequence Manager by selecting Sequence Manager on the BioPharma Finder home page.
- Select the Host Cell Protein Database Manager tab.
- The Host Cell Protein Database Manager page opens and displays the HCP Database table.
- Select the row in the table corresponding to the database you want to delete.
- The selected row is indicated by a blue arrow on the far left.
HCP Database table with row selected- Select Delete on the right side of the page.
- A confirmation dialog box appears, prompting you to confirm you want to delete database.
- Select Yes to permanently delete the database or select No to cancel.
- The application returns to the Host Cell Protein Database Manager page.