The BioPharma Finder application includes the Host Cell Protein (HCP) Database Manager to help detect and quantify trace host cell protein contaminants in biotherapeutics.
The application includes three default databases:
  • Chinese hamster Cricetulus griseus_uniprot_proteome_UP000001075
  • Human_uniprot_proteome_UP000005640
  • Mouse_uniprot_proteome_UP000000589
Three default HCP databases
Three default HCP databases
These default databases cannot be deleted. Custom databases can be added or deleted using the Host Cell Protein Database Manager.