Variable modifications are possible side-chain or terminal modifications whose specific sites and number of occurrences might not be known.
Upon installation, the BioPharma Finder application provides a default list of variable modifications, including N-glycans. You can access this list from the Protein Sequence Editor and set a sublist for quick loading into a protein sequence. You can also select which modifications will be visible in the modification editing panes and create custom modifications as necessary.