Remove static modifications - Remove static modifications - BioPharma Finder 5.2 Intact Mass Analysis User Guide - BioPharma Finder Software - Remove static modifications - BioPharma Finder Software - BioPharma Finder Software - topic
BioPharma Finder 5.2 Intact Mass Analysis User Guide
- Product Tree
- Ardia Platform > Software > BioPharma Finder Software
- Desktop Software > BioPharma Finder Software
- Document Type
- User Guide
- Software version
- 5.2
- In the Protein Sequence Map pane of the Protein Sequence Editor, position the cursor to the left of one of the following amino acid letter codes:
- For a side chain modification, the side chain residue of interest.
- For an N-terminal modification, the first residue of the chain.
- For a C-terminal modification, the last residue of the chain.
- Then, double-click.
- The Residue Properties and Modifications dialog box opens.
- The properties of the selected amino acid appear in the Residue Properties area. Check these properties to make sure that you selected the appropriate amino acid.
- (Optional) To globally remove a side chain modification applied to the selected amino acid letter, do one of the following:
- Select the Apply to All checkbox in the Side Chain Modification area and then select Clear.
- Select None in the Side Chain, N-Terminal, or C-Terminal Modification area (or in multiple areas).
- You can use the Apply to All checkbox to globally make changes only for side chain modifications.
- If you want to modify N- or C-terminal modifications, you must modify each instance individually.
- Select OK.
- When you remove a static modification, the blue highlight for the modified amino acid disappears in the Protein Sequence Map.
- If you selected Apply to All for a side chain modification, all of the blue highlights for the modified amino acids disappear.
- Select Save or Save as New in the command bar to save the modifications. Or, if you do not want to save the modifications to the sequence, select Cancel.