Static modifications are side-chain or terminal modifications that you can apply to a single site or all sites for a specified residue or terminal. These modifications can be made to the side chains, C-terminal, or N-terminal of a sequence.