The following table describes the columns in the table in the Disulfide Link Definitions pane of the Protein Sequence Editor.
Column | Description |
---|---|
Chain Number | Displays the number of the chain that includes the first cysteine (C) in the disulfide bond. |
Amino Acid Site Index | Displays the location within the chain of the first cysteine (C) in the disulfide bond. |
Chain Number | Displays the number of the chain that includes the second cysteine (C) in the disulfide bond. |
Amino Acid Site Index | Displays the location within the chain of the second cysteine (C) in the disulfide bond. |
Delete (button) | Deletes the disulfide bond corresponding to the selected row in the table in the Disulfide Link Definitions pane. |