Save a protein or peptide sequence under the same name (overwrite) - Save a protein or peptide sequence under the same name (overwrite) - BioPharma Finder 5.2 Intact Mass Analysis User Guide - BioPharma Finder Software - Save a protein or peptide sequence under the same name (overwrite) - BioPharma Finder Software - BioPharma Finder Software - topic
BioPharma Finder 5.2 Intact Mass Analysis User Guide
- Product Tree
- Desktop Software > BioPharma Finder Software
- Ardia Platform > Software > BioPharma Finder Software
- Document Type
- User Guide
- Software version
- 5.2
- In the Protein Sequence Editor, select Save in the command bar.
- If the sequence already exists, the following warning prompt appears: "The sequence name already exists and will be overwritten."
- Select Yes to have the current sequence overwrite previously saved data.
- Information from the saved sequence populates the columns of the sequence table on the Sequence Manager page.