The following table describes the parameters of the Modification Editor pane of the Protein Sequence Editor.
Parameter | Description |
---|---|
Modifications | Lists the modifications available to apply to an N-terminal, C-terminal, or side chain. |
N Terminal | Displays information about the N-terminal modification that you selected from the list. |
Mono. Mass | Displays the monoisotopic mass of the selected N-terminal modification. |
Avg. Mass | Displays the average mass of the selected N-terminal modification. |
Add | Opens the Add New Modification dialog box, so that you can add a custom N-terminal modification. |
Delete | Permanently removes the selected custom modification from the list of N-terminal modifications. |
C Terminal | Displays information about the C-terminal modification that you selected from the list. |
Mono. Mass | Displays the monoisotopic mass of the selected C-terminal modification. |
Avg. Mass | Displays the average mass of the selected C-terminal modification. |
Add | Opens the Add New Modification dialog box, so that you can add a custom C-terminal modification. |
Delete | Permanently removes the selected custom modification from the list of C-terminal modifications. |
Side Chain | Displays information about the side chain modification that you selected from the list. |
Mono. Mass | Displays the monoisotopic mass of the selected side chain modification. |
Avg. Mass | Displays the average mass of the selected side chain modification. |
Residues | Displays the amino acid residue or residues for the selected side chain modification. |
Add | Opens the Add New Modification dialog box, so that you can add a custom side chain modification. In this version of the dialog box, the Residues box is enabled so that you can specify specific residues for the modification. |
Delete | Permanently removes the selected custom modification from the list of side chain modifications. |