To create a new protein sequence or edit an existing protein sequence, you can do any of the following:
- In the Sequence Manager, you can import an existing BioPharma sequence file (.sequencebpf), and then open the sequence in the Protein Sequence Editor for editing.
- Open the Protein Sequence Editor from the Sequence Manager and then import a FASTA file that contains the protein sequence.
- Open the Protein Sequence Editor from the Sequence Manager and then manually enter the sequence.
After you enter the sequence and select its category, you can add static modifications and assign variable modifications to search against.