You can copy the content of the following panes to the clipboard for the types of BioPharma Finder analyses. You can then paste the copied content into a third-party application.
Copy commands for data results by type of analysis and pane
Command
Page/Pane
Analysis
Peptide/Oligo
Intact
Top Down
Copy
Process and Review page
  • Chromatogram pane
  • Peptide or Oligonucleotide Sequence Coverage pane
  • Full Scan Spectra pane
  • MS2 Spectra pane
Yes
Copy
Coverage page
  • Chromatogram pane
  • Protein or Oligonucleotide Sequence Coverage Map pane
Yes
Copy
Modification Summary page
  • Chromatogram pane
  • Peptide or Oligonucleotide Sequence Coverage pane
  • Full Scan Spectra pane
  • MS2 Spectra pane
Yes
Copy, Copy as Displayed, Copy per Global Settings
Process and Review page
  • Chromatogram pane
Yes
Yes
  • Deconvoluted Spectrum pane
Yes
  • Intact Fragmentation Deconvoluted Spectrum and Intact Deconvolution Deconvoluted Spectrum pane
Yes
  • Source Spectrum pane
Yes
  • Intact Fragmentation Source Spectrum and Intact Deconvolution Source Spectrum pane
Yes
Copy Data
Process and Review page
  • Deconvoluted Spectrum pane
Yes
  • Intact Fragmentation Deconvoluted Spectrum and Intact Deconvolution Deconvoluted Spectrum pane
Yes
Copy as Displayed, Copy per Global Settings
Spectra Comparison page | Mirror Plot pane
Yes
Copy
Process and Review page | ProSightBP Fragment Map pane
Yes