If you modify any processing method parameters, you must save the method to store the modified values for processing.
NOTE
You cannot overwrite a default processing method. You must save changes made to a default method under a new name.
All experiment names, sequence names, and method names are case-insensitive.
For more information on editing processing methods, refer to the workflow-specific "Working with a [workflow] processing method" chapters later in the User Guide.