Procedure
  1. On the BioPharma Finder home page, select the Peptide Mapping Analysis, Oligonucleotide Analysis, Intact Mass Analysis, or Top Down Analysis workflow link in the Experiment Types pane or below the splash graphic.
  2. The corresponding workflow page opens.
  3. In the Processing Method table, select the row (not the checkbox) for the processing methods you want to delete.
  4. The application highlights the selected method in blue.
  5. You can select only one method at a time to delete.
  6. Press the DELETE key and select Yes in the confirmation box.
  7. In the table in the Processing Method area, select a method row.
  8. NOTE
    You cannot delete any default processing method.