Change the location of a raw data file - Change the location of a raw data file - BioPharma Finder 5.2 Intact Mass Analysis User Guide - BioPharma Finder Software - Change the location of a raw data file - BioPharma Finder Software - BioPharma Finder Software - topic
BioPharma Finder 5.2 Intact Mass Analysis User Guide
- Product Tree
- Ardia Platform > Software > BioPharma Finder Software
- Desktop Software > BioPharma Finder Software
- Document Type
- User Guide
- Software version
- 5.2
- Open the Results page from the Load Results page or the Queue page.
- For more information on how to open the Results page, see the appropriate "Viewing [workflow name] results" topic in the User Guide specific to the workflow you are using.
- If the correct raw data files are in the most recent location matching the database, the application opens the experiment.
- If the raw data files are not in the most recent location, the application opens a dialog box where you can locate the raw data files in their new location. After locating the raw data files, select the files and then select Open.
- The default file location for raw data files is
C:\Xcalibur\data. - The application checks to see if the selected files match the files used in the experiment.
- If the files match, the application opens the experiment.
- If the files do not match, the application displays an error message and prompts the user to respecify the raw data file location.