The BioPharma Finder application requires one or more raw data files as input for every experiment, no matter the workflow. The raw data files contain your experimental data and are processed by the application according to the processing method you select.
NOTE
The larger the number and file size of raw data files you load, the longer the application might take to process the results.
Because the application repeatedly accesses the raw data files during data processing, your processing time might be very slow if the application accesses the raw data files through a network.
Copy the raw data files to your local computer for faster processing speed