You must disconnect from the Ardia platform in order to switch to local mode.
NOTE
If you have jobs processing in the run queue, check the statuses of the experiments before disconnecting from the Ardia platform. You cannot disconnect from the platform if you have experiments in the Running status. Stop the run queue or wait for the experiment to complete before disconnecting.
Procedure
  1. On the BioPharma Finder home page, select Help > Ardia Platform Management > Disconnect.
  2. Select OK.
  3. The application restarts in local mode.