When you open the Help menu from the BioPharma Finder home page while connected to the Ardia platform, you can access the following commands to perform basic platform management tasks:
- Register with the Ardia platform: Opens the Ardia Platform Registration dialog box.
- Link to Ardia platform: Links the user to the Ardia platform application.
- Connect: (Inactive when already connected to the Ardia platform)
- Disconnect: Disconnects the BioPharma Finder application from the Ardia platform and restarts the application in local mode.
- Deregister with the Ardia Platform: Deregisters the user from the Ardia platform.
- Manual Lock: Locks the BioPharma Finder application.
- Data Space Optimization: Removes temporary BioPharma Finder processing files to free up hard disk space.