NOTE
Thermo Fisher Scientific recommends using Rufus, which is a program for creating bootable USB drives, that can be downloaded here https://rufus.ie/en/.
Procedure
- Insert the USB drive into the computer.
- Run Rufus.
- In Rufus, configure the following settings under the Drive Properties area:
- Device: Use the drop-down to select the inserted USB drive.
- Boot selection: With the option button set to SELECT, browse for the required iso file.
- Partition scheme: Use the drop-down to select MBR.
- Target system: Use the drop-down to select BIOS or UEFI.
- Keep all default settings under the Format Options area.
- Select START.
- Keep all default options for any pop-up windows that appear and enter the default selection (OK and Yes).
- Wait until the Status area displays 100% READY.
- Select CLOSE and eject the USB drive from the computer.