Jump to main content
Features
This release includes the following new features and enhancements:
Enhancements
-
Updated the user interface (UI) and navigation to enhance intuitiveness and control, with clearer feature access and improved visibility into the Platform Link service status and connectivity.
-
Windows-level notifications provide information on key events, including APL service health, operational status, and registration updates.
-
Detailed messaging on the Ardia Platform Link home and registration screens informs the user of issues, including:
-
Instrument PC time not in sync with the server.
-
RabbitMQ service status notification.
-
Ardia Platform Link installs new versions of RabbitMQ version 4.1.0 and Erlang version 27.3.3 to improve reliability and performance.