Known defects
The following table lists known defects in the software with a brief abstract about the defect and information related to each defect’s severity and risk. The Item ID corresponds to the internal number assigned to each issue. Product management assesses the risk, which might differ significantly from the reported severity.
|
Software Section |
Item ID |
Abstract |
Severity |
Risk |
|---|---|---|---|---|
|
Ardia Platform Link |
756697 |
The instrument PC may require a reboot when re-validated with a URL different from the one used during initial registration. |
Low |
Low |
|
Ardia Platform Link |
756698 |
If the same device is configured on two different instrument PCs and is deleted from the configuration of one, the remaining PC with the device must be rebooted in order to maintain connection with the Ardia Platform. |
Medium |
Medium |