Jump to main content
Installation
Users can download the Ardia_Platform_Link_Setup_1.2.x.x.exe file and the Ardia Platform Link Setup Guide from the Thermo Fisher Scientific Digital Science Support (DSS) Resource Center: https://support.thermoinformatics.com/downloads/default.aspx
When installing the Ardia Platform Link application on an instrument PC, install it only after the Ardia Platform server is installed and running.