Known limitations
The following table lists the known limitations in the software, which are categorized by software section. Each row includes a brief abstract about the limitation.
Software Section
Abstract
Ardia Platform Link
If the instrument PC time is not synchronized with the time server, instruments might not register properly on the Ardia Platform.
Ardia Platform Link
Orbitrap Tribrid™ Series instruments require ICSW 4.2 QF1 patch or later for full compatibility with the Ardia Instruments application. Earlier versions are not fully supported.
Ardia Platform Link
Orbitrap Exploris™ Series instruments require ICSW 4.4 or later for full compatibility with the Ardia Instruments application. Earlier versions are not fully supported.