The following table lists known limitations in the software, which are categorized by software section. Each defect row includes a brief abstract about the limitation.
Software Section | Abstract |
|---|---|
Ardia Platform Link | If the instrument PC's time is not synchronized with the time server, instruments might fail to register properly on the Ardia platform. |
Ardia Platform Link | Orbitrap Tribrid™ Series instrument models require ICSW 4.2 QF1 patch or higher for full compatibility with the Instruments application. Lower versions are not fully supported. |
Ardia Platform Link | Orbitrap Exploris Series instrument models require ICSW 4.4 or higher for full compatibility with the Instruments application. Lower versions are not fully supported. |