The following table lists defects that were resolved between Ardia Platform Link Application 1.0 and 1.1 releases. The table excludes Help issues and cosmetic fixes, and some abstracts have been updated from their original versions to better describe the reported issues.
Software Section | Item ID | Abstract |
|---|---|---|
Ardia Platform Link | 402801 | After restarting the Ardia Platform Link services, the active system card might not display the chromatogram and the remaining sequence time. In this case, restart the services again. |
Ardia Platform Link | 465633 | Instruments show "Connection lost" due to RabbitMQ™ crash. |
Ardia Platform Link | 467055 | Excessive log file sizes for Ardia Platform Link services. |
Ardia Platform Link | 490002 | Change in IP address for Instrument PC causes connection loss until Ardia Platform Link services are restarted. |
Ardia Platform Link | 612852, 493689 | The Thermo Scientific TraceFinder™ application becomes unresponsive after installing the Ardia Platform Link application. |
Ardia Platform Link | 418136 | CPU usage increases after submitting a large number of injections in the Chromeleon CDS. |