Ardia Platform Link 1.1 is compatible with the Xcalibur™ 4.7 software and Chromeleon™ 7.3.2 MUc Chromatography Data System (CDS).
Refer to the system requirements for operating the Xcalibur™ software or Chromeleon™ CDS.
Before users can begin installing the Ardia Platform Link, the following requirement must be fulfilled:
  • An Ardia Platform account registered under the primary user is required to establish a connection to the Ardia Platform and perform the Ardia Platform URL validation.
IMPORTANT
The Ardia Core Software 1.1 release is a prerequisite for installing or upgrading to the Ardia Platform Link 1.1 application.
Ardia Platform Link 1.0 is not compatible with Ardia Core Software 1.1. Existing users must upgrade to Ardia Platform Link 1.1 to continue using the Ardia Instruments 1.1 application.