New features - Ardia Platform Link - Ardia Platform Link 1.1 Release Notes - Ardia Platform Link 1.1 Release Notes - New features - Features - Ardia Platform Link
Ardia Platform Link 1.1 Release Notes
- Product Tree
- Ardia Platform > External Ardia Tools > Ardia Platform Link
- Document Type
- Release Notes
- Software version
- 1.1
- The Ardia Platform Link Setup installs the following two new Windows services:
- Device Performance Metric
- Instrument Discovery Service
- The Ardia Platform Link application now includes an embedded browser window to facilitate connecting the Instrument PC to the Ardia Platform.
- The Ardia Platform Link application can now be upgraded to newer versions using the Ardia Platform Link Setup.
- The Ardia Audit Trail Viewer utility records an event for Ardia Platform Link registration.