This release includes these features:
- The installation of Ardia Platform Link installs the following Windows services:
- Device Integrator Service
- Device Registration Service
- Instrument Service Facade
- Local Device Service
- Proxy Client
- Users can connect the instrument PC to the Ardia Platform either during the installation of the Ardia Platform Link or later within the application.
- Ardia Platform Link can generate an Instrument Application Programming Interface (iAPI) license to control the usage of iAPI. With iAPI, users can create custom C# applications to modify instrument parameters in real-time.