Users can download the Ardia_Platform_Link_Setup.exe file and the Ardia Platform Link Setup Guide from the Digital Science Support (DSS) Resource Center:
https://support.thermoinformatics.com/downloads/default.aspx
When installing the Ardia Platform Link on an Instrument PC, do so only after the Ardia Platform server is installed and running.