The following table lists the known defect in the software. The defect row includes a brief abstract about the defect and information related to the defect's severity and risk. The Item ID corresponds to the internal number assigned to each issue. Product management assesses the risk based on the criteria listed above, which can differ significantly from the reported severity.
Software Section
Item ID
Abstract
Severity
Risk
Ardia Platform Link
402801
After restarting the Ardia Platform Link services, the active system card might not display the chromatogram and the remaining sequence time. In this case, restart the services again.
Low
Low