The Thermo Scientific™ Ardia™ Platform Data Sync application allows you to download raw data files directly from the Ardia Platform to your local Windows™ computer. Ardia Platform Data Sync reproduces the folder structure of the selected Ardia Platform folder, as shown in the following figures.
Ardia Platform Data Sync works by doing the following:
  1. Identifies all raw data files in a selected Ardia Platform folder.
  2. Adds each identified raw data file to the download queue.
  3. Downloads all files in the download queue, where the most recently modified file is downloaded last.
  4. NOTE
    If an injection was downloaded in a previous scan, it is ignored in all subsequent scans.
  5. NOTE
    Moving a folder on Ardia Platform does not change its modification date.
  6. Saves the downloaded files to the local folder that was selected as the download location.
  7. NOTE
    If a file with the same name already exists in the target location, Ardia Platform Data Sync adds a suffix (a number) to the name of the new file (for example, file_02.raw). If the suffix exceeds 100, the download is canceled.
  8. Scans for new files continuously and downloads new raw files whenever they are uploaded to the Ardia Platform (if configured)
For more information on setting up and using Ardia Platform Data Sync, see these topics: