Ardia Platform Data Sync has three pages (Download, Queue, and Finished), a Settings icon, and a Help icon. On the Download page, you can select which folders you want to download from the Ardia Platform onto your local computer. The Queue page contains a list of downloads that are currently in progress or queued. The Finished page contains a list of all completed and failed downloads. The following figures show these pages.
| No. | Description | No. | Description |
|---|---|---|---|
1 | View the current download selections | 6 | Start the download |
2 | View in-progress downloads | 7 | View available storage |
3 | View completed and failed downloads | 8 | Select a local download location |
4 | View the version number, legal notices, EULA, and user guide | 9 | Select Ardia Platform folders to download |
5 | Configure additional settings |
| No. | Description | No. | Description |
|---|---|---|---|
1 | View a selected file's summary | 3 | Remove the selected file from the download queue |
2 | Open the local folder where the raw file is saved | 4 | Stop an ongoing download |
| No. | Description | No. | Description |
|---|---|---|---|
1 | View a selected file's summary | 3 | Re-download a file that previously failed |
2 | Open the local folder where the raw file is saved | 4 | Stop an ongoing download |
The following table describes the statuses that appear on the Queue page and the Finished page.
Status | Description |
|---|---|
Queue page | |
Queued | The download is planned but has not started yet. |
Downloading | The file is being actively downloaded. |
Re-Queued | A previous download attempt failed and will be restarted later. This might happen for the following reasons:
|
Finished page | |
Finished | The download was successful. |
Error | The download has failed. This might happen for the following reasons:
You can select a failed download to display its summary, which includes a status message that explains the error. |
Canceled | The download was canceled from the Queue page. |
The following table describes the tasks and procedures that can be performed from each of Ardia Platform Data Sync's pages.
Procedure
- To use Ardia Platform Data Sync, perform any of the tasks in the following table.
Task | |
|---|---|
Download page | |
Select the Ardia Platform folders to download | Procedure
|
Set a specific date for Ardia Platform Data Sync to scan an Ardia Platform folder for new files | Procedure
|
Select a download location on your computer | Procedure
|
Start a download | Procedure
|
Stop a download | Procedure
|
Queue page | |
View a file's summary | Procedure
|
Cancel an individual download | Procedure
|
Finished page | |
View a file's summary | Procedure
|
Retry an individual download | Procedure
|
Settings page | |
Select how often Ardia Platform Data Sync automatically scans the Ardia Platform folders for new files | Procedure
|
Sign out of Ardia Platform Data Sync | Procedure
|
Reset Ardia Platform Data Sync | Procedure
|