The following table lists the known defect in the software. The defect row includes a brief abstract about the defect and information related to the defect's severity and risk. The Item ID corresponds to the internal number assigned to each issue. Product management assesses the risk based on the criteria listed above, which can differ significantly from the reported severity.
Software Section | Item ID | Abstract | Severity | Risk |
|---|---|---|---|---|
Ardia Platform Data Sync | 434576 | If a user gains access to an Ardia Platform subfolder while Ardia Platform Data Sync is downloading data, the raw files in the subfolder are not downloaded in the next scan. To download the files, users must remove and re-select the Ardia Platform folder from the Download page. | Medium | Low |