The following table describes the Ardia Platform Data Sync (ADS) application versions and their compatibility with different versions of the Ardia Core software.
| Ardia Core software version 1.0 | Ardia Core software version 1.1 |
|---|---|---|
ADS version 1.0 | Compatible | Not Compatible |
ADS version 1.1 | Not Compatible Upgrade to Ardia Core software version 1.1. | Compatible |
IMPORTANT
Thermo Fisher Scientific recommends upgrading to the Ardia Core software version 1.1 before upgrading your Ardia Platform Data Sync application.