The Queue page of the Ardia Platform Data Sync application allows you to view all in progress or pending sync jobs.
Procedure
- Select the Queue tab.
- To view an in progress or queued file’s summary, select a data file from the list.
- The Summary pane opens.
- (Optional) Select Open Local Folder to navigate directly to the file on your PC.
- The Windows file explorer dialog box opens.
- (Optional) Select Cancel Syncing to cancel an individual sync.
- The selected file is removed from the queue and any pending syncs in the queue are paused.
- (Optional) Select Start Sync to sync the remaining files in the queue.
- (Optional) Select Stop to stop the in-progress sync job.