You can remove the Ardia Platform Data Sync application from your PC.
Procedure
  1. Do one of the following to start the uninstallation process:
  2. Double-click the original Thermo Scientific Ardia Platform Data Sync.exe file saved to your PC and, if prompted, select Yes to allow the application to make changes to your device.
  3. NOTE
    You might need administrative privileges on your PC to run the Ardia Platform Data Sync Setup.
  4. Select Control Panel > Programs and Features, then double-click Thermo Scientific Ardia Platform Data Sync.
  5. Select Uninstall in the setup or select Yes in the Thermo Scientific Ardia Platform Data Sync dialog box to uninstall the application.
  6. The setup displays the uninstallation progress and a confirmation message upon completion.
  7. (Optional) When the uninstallation is complete, select Log to view the setup summary log file.
  8. NOTE
    The IQ reports are saved to C:\Program Files\Thermo Scientific\Ardia\ADS.
    Logs are saved to C:\ProgramData\Thermo Scientific\Ardia\ADS.
  9. Select Close to exit the setup.
  10. The Ardia Platform Data Sync application is removed from your PC.