The Thermo Scientific™ Ardia™ Platform Data Sync application allows you to sync data files between the Ardia Platform and your local PC. With this application, you can transfer data in the following ways:
- From your local PC to the Ardia Repository
- From the Ardia Repository to your local PC
The Ardia Platform Data Sync application reproduces the folder structure of the selected source and destination folders, as shown in the following figures, and syncs all unique subfolders and files in the selected directories.
For details on each page of the application and how the application syncs data, see these topics:
To set up and begin using the Ardia Platform Data Sync application, see these topics: