The Ardia Platform Data Sync Setup installs the Ardia Platform Data Sync application, enabling file syncing between the Ardia Repository and your local PC.
IMPORTANT
For enhanced security and protection of your personal information, it is recommended to use dedicated, personal devices and maintain strict control over your access credentials. If shared devices are unavoidable, consider implementing additional security measures such as token expiration and multi-factor authentication to mitigate risks.
Procedure
  1. Download the Thermo Scientific Ardia Platform Data Sync Setup installer to your PC.
  2. NOTE
    The Ardia Platform Data Sync installer can be downloaded from the Thermo Fisher Scientific Digital Science Support (DSS) Resource Center.
  3. Right-click the Thermo_Scientific_Ardia_Platform_Data_Sync.exe file and select Run as Administrator to open the setup wizard.
  4. If prompted, select Yes to allow the application to make changes to your instrument PC.
  5. The Thermo Scientific Ardia Platform Data Sync Setup opens.
  6. Review the license agreement and then select the I Agree to the License Terms and Conditions checkbox.
  7. Select Next.
  8. Review the system requirements, then select Install.
  9. The setup displays the installation progress and a confirmation message upon completion.
  10. (Optional) When the installation is complete, you can do the following in the setup:
  11. Select IQ Report to view the installation qualification (IQ) report.
  12. Select Log to view the setup summary log file.
  13. Select Open IQ Reports Folder to open the system folder containing the IQ report and log PDF files.
  14. NOTE
    The installation files, program data, application startup executable, IQ reports are saved to C:\Program Files\Thermo Scientific\Ardia\ADS.
    Logs are saved to C:\ProgramData\Thermo Scientific\Ardia\ADS.
  15. Select Close to exit the setup.
  16. The installation of the Ardia Platform Data Sync application is complete.
  17. Continue to Register the Ardia Platform Data Sync application on the Ardia Platform.