Enhancements - Enhancements - Ardia Platform Data Sync 1.1 Release Notes - Ardia Platform Data Sync - Ardia Platform Data Sync 1.1 Release Notes - Enhancements - Features - Ardia Platform Data Sync 1.1 Release Notes - Ardia Platform Data Sync - topic
Ardia Platform Data Sync 1.1 Release Notes
- Product Tree
- Ardia Platform > External Ardia Tools > Ardia Platform Data Sync
- Document Type
- Release Notes
- Software version
- 1.1
- Quickly register or unregister the Ardia Platform Data Sync application to the Ardia Platform.
- Customizable folder monitoring:
- Monitor up to 20 parent folders, including their subfolders, on a local PC, the Ardia Platform, or both for newly added data files.
- Automatically download or upload data files to their unique folders.