The following table lists known defects in the software with a brief abstract about the defect and information related to each defect’s severity and risk. The Item ID corresponds to the internal number assigned to each issue. Product management assesses the risk, which might differ significantly from the reported severity.
Software Section | Item ID | Abstract | Severity | Risk |
|---|---|---|---|---|
Ardia Platform Data Sync | 756217 | If a user gains access to an Ardia Platform subfolder while Ardia Platform Data Sync is downloading data, the raw files in the subfolder are not downloaded in the next scan. To download the files, users must remove and re-select the Ardia Platform folder from the Download page. | Medium | Low |
Ardia Platform Data Sync | 650904 | If the display scale is changed, the pick folder dialog box will not appear until the application is restarted. | Low | Low |
Ardia Platform Data Sync | 641944 | Files not listed as allowed in the Ardia Global Settings utility display an error indicating a failed malicious content scan, which might be misleading. | Low | Low |
Ardia Platform Data Sync | 753782 | Disk space error occurs when the server connection is lost. | Medium | Medium |
Ardia Platform Data Sync | 754244 | The .methodbpf file type does not upload, even when Biopharma Finder Method files are selected as allowed in the Ardia Global Settings utility. To resolve this, you must add .methodbpf as a custom file type. | Medium | Medium |
Ardia Platform Data Sync | 754258 | The .pdResultMeta file type does not upload, even when Proteome Discoverer Method files are selected as allowed in the Ardia Global Settings utility. Additionally, it cannot be added as a custom file type because it already exists in the list. | Medium | Medium |