Jump to main content
Technical
-
The runtime libraries of the Ardia Configuration Utility portable application are cached %localappdata%\Temp\.net\ArdiaConfigurationUtility.
-
Logs generated during operation of the Ardia Configuration Utility are stored on Windows computer at C:\ProgramData\ThermoFisher\ArdiaConfigurationUtility\Logs.