Configure NFS backup and archive for the Ardia Server
Use the Configure tab to set up an external Network File System (NFS) and enable backup and archive.
Procedure
-
In the Ardia Configuration Utility, select the Configure tab
The following options are available:
-
Cancel - Returns to the Console tab.
-
Apply - Disabled when there are no changes to the settings of either Backup or Archive. Enabled when a change is detected and applies the respective settings for both Backup and Archive.
-
-
Complete the following fields to configure the backup.
-
NFS Server IP Address - Enter the IP address of the NFS server.
-
Export Path - Enter the path to the file share (use appropriate syntax of forward slashes).
-
Version - Select the relevant version of NFS protocol for the chosen NFS storage device.
-
-
Complete the following fields to configure an archive.
-
NFS Server IP Address - Enter the IP address of the NFS server.
-
Export Path - Enter the path to the file share (use appropriate syntax of forward slashes).
-
Version - Select the relevant version of NFS protocol for the chosen NFS storage device
Note: The Backup and Archive fields automatically validate the entries and display prompts if any corrections are required.
Note: Pop-up messages warn about potential configuration issues or errors.
-