About the Instruments application
Ardia Instruments is an Ardia Core application that lets you manage, schedule, and monitor data acquisition on all instruments connected to the Ardia Platform. It provides centralized lab management tools with the features described in the following table.
|
Feature |
Description |
|---|---|
|
The Systems view lets you view the entire fleet of systems in your laboratory. This page serves as a dashboard that consists of cards. Each card represents a system (examples include LC, GC, IC, LC/MS, GC/MS or IC/MS). |
|
|
The Scheduler view integrates seamlessly with the Systems feature. Using the Scheduler, you can reserve an instrument based on its availability to plan maintenance or perform sample analysis. This eventually helps you track instrument usage and downtime. |
|
|
The Logbook view provides a way to record maintenance, calibration, diagnostics, software updates, and other operational events for your configured instruments. Here, you can view auto-populated events or create an event manually. |
|
|
The Request Support button lets you request technical support. Using this feature, you can request support by creating a service ticket. You can also share the relevant log files and other required files to assist service team. |