Minimum requirements
Ardia Instruments 1.1.1 is compatible with the Xcalibur™ 4.7 software and Chromeleon™ 7.3.2 MUc Chromatography Data System (CDS) or later versions of both.
Refer to the minimum requirements for using Ardia Core Software in the Ardia Core Software 1.2 Release Notes.