Known defects
The following table lists known defects in the software with a brief abstract about the defect and information related to each defect’s severity and risk. The Item ID corresponds to the internal number assigned to each issue. Product management assesses the risk, which might differ significantly from the reported severity.
|
Software section |
Item ID |
Abstract |
Severity |
Risk |
|---|---|---|---|---|
|
Systems |
365457 |
Sometimes, the System card does not display the complete chromatogram when it contains significantly delayed eluting peaks. |
Medium |
Medium |
|
Systems |
366238 |
The number of sequences displayed on the System card represents the total number of sequences in the queue (Chromeleon CDS acquisition). |
Low |
Low |
|
Systems |
381930 |
The System card for the disconnected instruments does not display the current or upcoming Scheduler reservations. |
Low |
Low |
|
Systems |
756689, 784305 |
User interface issues might occur after upgrading the Ardia Palform if the browser cache is not cleared. |
Low |
Low |
|
Systems |
756692 |
When the instrument configuration changes, initial auto-entries in the logbook might not display. |
Low |
Low |
|
Systems |
877752 |
The Thermo Scientific™ Orbitrap™ Ascend™ Tribrid™ mass spectrometer name and model are unknown, and the image is missing. |
Medium |
Low |