Known limitations
The following table lists the known limitations in the application, which are categorized by application section. Each row includes a brief abstract about the limitation.
Application section
Abstract
Systems
Chromeleon CDS users must save the instrument configuration in the software to ensure that the configuration details synchronize with the Ardia Instruments application.
Systems
Chromeleon CDS users can view the chromatogram on the System card only for the current default channel.
Systems
If a user connects the instrument PC to the Ardia Platform through the Ardia Platform Link application, it might take up to one to two minutes for the Waiting for Setup page to open.
Systems
The Thermo Scientific™ Orbitrap™ Tribrid™ mass spectrometer requires the ICSW 4.2 QF1 patch or later for full compatibility with the Instruments application. Earlier versions are not fully supported.
Systems
The Thermo Scientific™ Orbitrap™ Exploris™ mass spectrometer requires ICSW 4.4 or later for full compatibility with the Instruments application. Earlier versions are not fully supported.
Systems
The injection time remaining that is displayed on a System card might not be accurate when a method is used for the first time. Prediction accuracy will improve with repeated use of the method.
Systems
The Thermo Scientific™ Orbitrap™ Exploris™ mass spectrometer with ICSW 5.1 does not support auto-calibration entries recorded in the Logbook.