Ardia Instruments Application Quick Start Guide
This guide outlines the basic steps to connect your instruments to the Thermo Scientific™ Ardia™ Instruments application on the Ardia Platform. With the Instruments application, you can remotely manage, schedule, and monitor data acquisition on your instruments.
Before you can use the Instruments application, you first need to install and set up the Ardia Platform Link application on each instrument PC to facilitate communication between your instruments and the Ardia Platform. For more information, refer to the Ardia Platform Link Setup Guide and the Ardia Instruments Application User Guide.
Part 1: Install the Ardia Platform Link application
GUID-AABF8ED6-6138-4DE1-A086-A4D0564435F4-print.pngThe Ardia Platform Link application must be installed and set up on each instrument PC.
1. Install the Ardia Platform Link application
Run the Ardia_Platform_Link_Setup_1.2.x.x.exe file and select Install.
GUID-09D073F8-FF97-420B-BD80-970C4BA6BD1B-print.png
2. Register the Ardia Platform Link application
Enter the Ardia Platform URL and then select Register.
GUID-D64B0E52-6CF3-42F2-B0C5-4793A800D830-print.png
3. Enter your Ardia Platform credentials
Enter your Ardia Platform credentials to complete the registration and connection of your instrument PC to the Ardia Platform.
GUID-CB77AE04-A9B0-4920-926C-086CA65CBFFE-print.png
4. Verify the connection
The Ardia Platform URL status shows Connected. You can now view your system card in the Instruments application (see Set up your system in the following section).
GUID-492A032D-C303-4BA2-B191-81EE944E42C2-print.png
Part 2: Access the Instruments application and set up your system cards
GUID-AABF8ED6-6138-4DE1-A086-A4D0564435F4-print.pngNow that your instrument PC is connected to the Ardia Platform, you can view your system cards in the Instruments application.
1. Sign in to the Ardia Platform
Go to the Ardia Platform URL in a browser and sign in to the Ardia Platform.
GUID-DAE1DA64-42AC-414A-BF15-261185101B03-print.png
2. Launch the Ardia Instruments application
Open the Instruments application from the Ardia Platform Home page.
GUID-92519EC8-3E72-4FDA-8F94-DEC4F12BFD73-print.png
3. (Optional) Connect your lab to Thermo Fisher
Select Continue to connect. If you do not have permission, connect later from the Settings page.
GUID-BEE9D09C-A5EA-490A-8930-023A2AFF863F-print.png
4. Set up your system
On the Waiting for Setup view, select the Complete Setup option on the card of your instrument.
GUID-B1BF3ADA-0284-4CA1-A33D-B08F1C72D52C-print.png
5. Finish setup
Enter the required instrument details and select, and select Finish Setup.
GUID-717CF858-23C0-4FDC-8B13-36329FD17573-print.png
6. Start managing connected systems
On the Active view, view all your set up systems and manage your lab in one place.
GUID-B281A3B7-A2C9-4F59-8B05-101BEFFCDF2D-print.png
© 2026 Thermo Fisher Scientific Inc. All rights reserved.
All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
Pub. No. BRE0062367, Revision C
For Research Use Only. Not for use in diagnostic procedures.