The following table lists defects that were resolved between Instruments Application 1.0 and 1.1 releases.
The table excludes Help issues and cosmetic fixes, and some abstracts have been updated from their original versions to better describe the reported issues.
Software Section
Item ID
Abstract
Scheduler
375802
When the assignee and the initiator are the same user, the notifications are duplicated and sent twice.
Instrument connectivity
402801
After restarting the Ardia Platform Link services, the active system card might not display the chromatogram and the remaining sequence time. In this case, restart the services again.
Systems
424612
When a device is removed from the instrument configuration, the Instruments application fails to display the comparison window.
Instrument connectivity
343958
The creation of instruments with identical names within a Chromeleon CDS installation is not supported.
Systems
383258
The sequence information is not displayed on the System card when the queue is stopped after the injection or stopped after the sequence.
Systems
489963
In the Request Support workflow, the symptom list is not displayed for TSQ Quantis™ Plus Triple Quadrupole Mass Spectrometer.
Systems
451733, 494413
System cards might show inconsistent real-time sequence information.
Systems
492609
The Go to Scheduler link should be disabled (grayed out) when an instrument configuration change is detected.
Scheduler
487553
Scheduler notifications might still be sent to users even when their notification preference is disabled.
Systems/Scheduler
434403
Users may occasionally receive duplicate notifications.
Systems
606438
Real-time sequence information might stay in a repeated 20-second countdown loop, while the sequence continues to run.
Systems
365954
Negative chromatographic trace not displayed in mini plot view on system card.
Systems
426856
Request Support submission intermittently fails.
Systems
494263
When acquiring data in Xcalibur, injection name displayed on System card did not match the exact raw file name.
Systems
633412
Evosep One is not uniquely identified by its serial number; upgrade to the latest driver for full support and proper detection.