This guide outlines the basic steps to connect your instruments to the Thermo Scientific™ Ardia™ Instruments application on the Ardia Platform. With the Instruments application, you can remotely manage, schedule, and monitor data acquisition on your instruments.
Before you can use the Instruments application, you first need to install and set up the Ardia Platform Link application on each instrument PC to facilitate communication between your instruments and the Ardia Platform. For more information, refer to the Ardia Platform Link Setup Guide and the Ardia Instruments Application User Guide.
Part 1: Install the Ardia Platform Link application
1. Install the Ardia Platform Link application | 2. Register the Ardia Platform Link application |
3. Enter your Ardia Platform credentials | 4. Verify the connection |
Part 2: Access the Instruments application and set up your system cards
1. Sign in to the Ardia Platform | 2. Launch the Ardia Instruments application |
3. (Optional) Connect your lab to Thermo Fisher | 4. Set up your system |
5. Finish setup | 6. Start managing connected systems |
© 2024 Thermo Fisher Scientific Inc. All rights reserved.
All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
Pub. No. BRE0062367, Revision B
For Research Use Only. Not for use in diagnostic procedures.