The following table describes the options that allow users to setup of an instrument with an undefined configuration.
Function | Subfunction | Description |
|---|---|---|
Complete Setup | N/A | Adds or modifies the instrument attribute details and initiates real-time monitoring. |
Does the system include an MS? | N/A | Specifies if the undefined system includes an MS or does not include an MS. |
Define undefined instrument type(s) | Identify instrument type | Defines the undefined instrument types. |
NOTE
Continue with the remaining steps starting from System Setup function in the Waiting for Setup table.