The following table lists the common functions that are used for creating a group reservation.
Function | Subfunction | Description |
|---|---|---|
Back | N/A | Closes the Scheduler Assistant without saving any changes. |
Delete | N/A | Deletes an already existing group reservation. |
Date Range | N/A | See Date Range. |
Instrument pane | N/A | Displays the selected instruments from the Instrument page of the Scheduler Assistant. |
Add Instruments | Adds the instruments to the reservation. |